missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ruthenium(III) 2,4-pentanedionate, Ru 24% min., Alfa Aesar™

For Hydrogenation

Manufacturer:  Thermo Fisher Scientific 1056806

 View more versions of this product

Catalog No. AA1056806



Specifications

Specifications

Ruthenium(III) 2, 4-pentanedionate
14284-93-6
MFCD00000030
acetylacetone ruthenium iii salt, ruthenium acetylacetonate, ruthenium iii acetylacetonate, ruthenium, tris 2,4-pentanedionato-8ci, ruthenium, tris 2,4-pentanedionato-kappao,kappao'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-o,o'-, oc-6-11, tris 2,4-pentanedionato ruthenium iii, tris pentane-2,4-dionato-o,o' ruthenium, z-4-oxopent-2-en-2-olate; ruthenium 3+
RTZYCRSRNSTRGC-LNTINUHCSA-K
(Z)-4-oxopent-2-en-2-olate;ruthenium(3+)
5488829
Crystalline
260°C (decomposition)
Ru 24% min.
C15H21O6Ru
Red crystals
Soluble in most organic solvents such as acetone,chlorinated hydrocarbons,alcohols,cyclohexane and benzene. Insoluble in water.
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Ru+3]
398.397
398.4
White
5g

Chemical Identifiers

14284-93-6
398.397
RTZYCRSRNSTRGC-LNTINUHCSA-K
5488829
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Ru+3]
C15H21O6Ru
MFCD00000030
acetylacetone ruthenium iii salt, ruthenium acetylacetonate, ruthenium iii acetylacetonate, ruthenium, tris 2,4-pentanedionato-8ci, ruthenium, tris 2,4-pentanedionato-kappao,kappao'-, oc-6-11, ruthenium, tris 2,4-pentanedionato-o,o'-, oc-6-11, tris 2,4-pentanedionato ruthenium iii, tris pentane-2,4-dionato-o,o' ruthenium, z-4-oxopent-2-en-2-olate; ruthenium 3+
(Z)-4-oxopent-2-en-2-olate;ruthenium(3+)
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 238-193-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents