missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ruthenium black, 99.9% (metals basis), Thermo Scientific Chemicals
$531.84 - $1888.47
Chemical Identifiers
CAS | 7440-18-8 |
---|---|
Molecular Formula | Ru |
Molecular Weight (g/mol) | 101.07 |
MDL Number | MFCD00011207 MFCD03458417 |
InChI Key | KJTLSVCANCCWHF-UHFFFAOYSA-N |
Synonym | on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon |
PubChem CID | 23950 |
ChEBI | CHEBI:30682 |
IUPAC Name | ruthenium |
SMILES | [Ru] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-18-8 | |
101.07 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
23950 | |
ruthenium |
Ru | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
CHEBI:30682 | |
[Ru] |
Specifications
7440-18-8 | |
Ru | |
UN3089 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium | |
23950 | |
99.9% | |
Ruthenium black |
2 g | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
[Ru] | |
101.07 | |
CHEBI:30682 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228-H500
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 231-127-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only