Please login to your online account to display your discounted pricing

Ruthenium on activated carbon, 5% Ru, unreduced, ca. 50% moisture, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  195460250

 View more versions of this product

Catalog No. AC195460250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Ruthenium on activated carbon
Quantity 25g
Name Note 5% Ruthenium
CAS 7440-18-8
CAS 7440-44-0
CAS 7732-18-5
Density 1.02
Physical Form Powder
Assay 5% Ru on dry basis (typically)
Molecular Formula Ru
Formula Weight 101.07
MDL Number MFCD00011207
Fieser 01,983; 05,574
Merck Index 15, 8433
Packaging Glass bottle
Surface Area 900 m2/g (BET) (typically)
Color Black

Specifications

Chemical Name or Material Ruthenium on activated carbon
Name Note 5% Ruthenium
CAS 7440-18-8
CAS 7440-44-0
CAS 7732-18-5
Assay 5% Ru on dry basis (typically)
Molecular Formula Ru
Formula Weight 101.07
MDL Number MFCD00011207