Please login to your online account to display your discounted pricing

Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  198100010

 View more versions of this product

Catalog No. AC198100010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 101.07
Quantity 1g
Color Gray
Merck Index 15, 8433
Purity Grade For trace metal basis analysis
Assay Percent Range 99.90%
Molecular Formula Ru
Chemical Name or Material Ruthenium
Name Note -200 mesh
CAS 7440-18-8
Health Hazard 2 GHS H Statement:
Flammable solid.
MDL Number MFCD00011207
Packaging Glass bottle
Total Trace Metal Impurities 0.1% max.
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ruthenium
Name Note -200 mesh
CAS 7440-18-8
Purity Grade For trace metal basis analysis
Physical Form Powder
Assay Percent Range 99.90%
Molecular Formula Ru
Formula Weight 101.07
MDL Number MFCD00011207
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.

SDS & Safety Documents