Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium, 5% on carbon paste, D101002-5

Chemical Identifiers

Chemical Name or Material Ruthenium
CAS 7440-18-8
Name Note 5% on carbon paste, D101002-5
MDL Number MFCD03458417
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3685006

  SDS
alfa aesar™
H3685006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3685014

  SDS
alfa aesar™
H3685014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3685022

  SDS
alfa aesar™
H3685022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Ruthenium
Name Note 5% on carbon paste, D101002-5
CAS 7440-18-8
TSCA TSCA
Physical Form Solid
MDL Number MFCD03458417
Odor Odorless
Color Dark gray
Quantity 5g