Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium, 5% on alumina, D302011-5

Chemical Identifiers

Chemical Name or Material Ruthenium
CAS 7440-18-8
Name Note 5% on alumina, D302011-5
MDL Number MFCD03458417
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3788806

 
alfa aesar™
H3788806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3788814

 
alfa aesar™
H3788814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3788822

 
alfa aesar™
H3788822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Ruthenium
TSCA TSCA
Odor Odorless
Name Note 5% on alumina, D302011-5
Physical Form Powder
MDL Number MFCD03458417
CAS 7440-18-8
Color Dark gray
Quantity 5g