missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet

Manufacturer:  Alfa Aesar™ 4433822

 View more versions of this product

Catalog No. AA4433822



Specifications

Specifications

Ruthenium
7440-18-8
MFCD03458417
Insoluble in water.
[Ru]
101.07
CHEBI:30682
Odorless
5% on activated carbon powder, Reduced, nominally 50% water wet
Ru
7ui0tkc3u5
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
Powder
100g

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD03458417
7ui0tkc3u5
CHEBI:30682
[Ru]
Safety and Handling

Safety and Handling

EINECSNumber : 231-127-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents