Learn More
Ruthenium, 5% on activated carbon powder, reduced, Thermo Scientific Chemicals
Aromatic ring hydrogenation
Supplier: Thermo Scientific Chemicals 01174822
Description
It is used in hydrogenation of aromatic ring systems, particularly the hindered systems. It finds wide use for the reduction of aliphatic carbonyl compounds and carboxylic acids to alcohols, hydrogenation of anilines to cyclohexylamines, synthesis of ammonia, depolymerization of carbohydrates, Fischer-Tropsch synthesis, and hydrogenation of glucose to produce sorbitol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ruthenium | |
7440-18-8 | |
Ru | |
UN1362 | |
Insoluble in water. | |
[Ru] | |
101.07 | |
CHEBI:30682 |
5% on activated carbon powder, Reduced | |
100 g | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium | |
23950 | |
Odorless |
Chemical Identifiers
7440-18-8 | |
101.07 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
23950 | |
ruthenium |
Ru | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
CHEBI:30682 | |
[Ru] |
Safety and Handling
GHS H Statement
H228-H252-H315-H319-H335
Flammable solid.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235+P410-P240-P241-P280-P370+P378q
H228-H252
DOTInformation : Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: CARBON, ACTIVATED
EINECSNumber : 231-127-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only