Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium, 5% on activated carbon paste, Type 619

Chemical Identifiers

Chemical Name or Material Ruthenium
CAS 7440-18-8
Name Note 5% on activated carbon paste, Type 619
MDL Number MFCD03458417
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3600406

  SDS
alfa aesar™
H3600406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3600414

  SDS
alfa aesar™
H3600414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3600422

  SDS
alfa aesar™
H3600422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Ruthenium
Color Dark gray
CAS 7440-18-8
Name Note 5% on activated carbon paste, Type 619
TSCA TSCA
MDL Number MFCD03458417
Odor Odorless
Quantity 5g
Physical Form Powder