missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium, 5% on activated carbon paste, Type 619

Manufacturer:  Alfa Aesar™ H3600414

 View more versions of this product

Catalog No. AAH3600414



Specifications

Specifications

Ruthenium
7440-18-8
MFCD03458417
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
Powder
Gray
5% on activated carbon paste, Type 619
Ru
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
[Ru]
101.07
CHEBI:30682
Odorless
25g

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents