missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium, 4% on 6.35mm (0.25in) alumina rings

Manufacturer:  Alfa Aesar™ 4459336

 View more versions of this product

Catalog No. AA4459336



Specifications

Specifications

Ruthenium
7440-18-8
MFCD00011207
Insoluble in water.
[Ru]
101.07
CHEBI:30682
500g
4% on alumina rings, 6.35mm (0.25 in.)
Ru
7ui0tkc3u5
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
Gray-White

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207
7ui0tkc3u5
CHEBI:30682
[Ru]
Safety and Handling

Safety and Handling

EINECSNumber : 231-127-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents