missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium, 2% on 3.18mm (0.125in) alumina pellets

Manufacturer:  Alfa Aesar™ 4457522

 View more versions of this product

Catalog No. AA4457522



Specifications

Specifications

Ruthenium
100g
7440-18-8
MFCD00011207
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
S.A. nominally 200m
Odorless
2% on alumina pellets, 3.18mm (0.125 in.)
Ru
7ui0tkc3u5
[Ru]
101.07
CHEBI:30682
nominally 200m2/g

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207
7ui0tkc3u5
CHEBI:30682
[Ru]
Safety and Handling

Safety and Handling

EINECSNumber : 231-127-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents