missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ruthenium, 2% on 3.18mm (0.125in) alumina pellets

Manufacturer:  Alfa Aesar™ 4457514

 View more versions of this product

Catalog No. AA4457514



Specifications

Specifications

Ruthenium
7440-18-8
MFCD00011207
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
Odorless
25g
2% on alumina pellets, 3.18mm (0.125 in.)
Ru
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
[Ru]
101.07
CHEBI:30682
nominally 200m2/g

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents