missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rutaecarpine 98.0+%, TCI America™
Supplier: TCI America R0102200MG
Specifications
Rutaecarpine | |
262°C | |
C18H13N3O | |
200 mg | |
ACVGWSKVRYFWRP-UHFFFAOYSA-N | |
3,13,21-triazapentacyclo[11.8.0.0²,¹â°.0â´,â¹.0¹âµ,²â°]henicosa-1(21),2(10),4,6,8,15,17,19-octaen-14-one | |
65752 | |
287.32 | |
Crystalline Powder |
84-26-4 | |
Yellow | |
MFCD00210551 | |
8,13-Dihydroindolo[2′C,3′C:3,4]pyrido[2,1-b]quinazolin-5(7H)-one | |
O=C1N2CCC3=C(NC4=CC=CC=C34)C2=NC2=CC=CC=C12 | |
287.32 | |
CHEBI:8922 | |
≥98.0% (HPLC,N) |
Chemical Identifiers
84-26-4 | |
287.32 | |
ACVGWSKVRYFWRP-UHFFFAOYSA-N | |
65752 | |
3,13,21-triazapentacyclo[11.8.0.0²,¹â°.0â´,â¹.0¹âµ,²â°]henicosa-1(21),2(10),4,6,8,15,17,19-octaen-14-one |
C18H13N3O | |
MFCD00210551 | |
8,13-Dihydroindolo[2′C,3′C:3,4]pyrido[2,1-b]quinazolin-5(7H)-one | |
CHEBI:8922 | |
O=C1N2CCC3=C(NC4=CC=CC=C34)C2=NC2=CC=CC=C12 |
Safety and Handling
RTECSNumber : NM3454000
TSCA : No
Recommended Storage : Refrigerator