missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Runx3 Mouse anti-Human, eFluor 660, Clone: R3-5G4, eBioscience™

Mouse Monoclonal Antibody

$160.95 - $397.30

Specifications

Antigen Runx3
Clone R3-5G4
Host Species Murine
Gene Alias AML2, CBFA3
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124555 Life Technologies
LS50981780
25μg Each for $160.95
501124556 Life Technologies
LS50981782
100μg Each for $397.30
Specifications

Description

This R3-5G4 monoclonal antibody recognizes human Runx3, a 44kDa transcription factor that belongs to the Runt domain-containing family. Runx3 is expressed in hematopoietic cells such as T cells, B cells, and myeloid cells. This transcription factor is also expressed in non-hematopoietic tissues, including the epidermis, bone, and dorsal root ganglia. Runx3 has been established to play a critical role in thymocyte development. More specifically, Runx3 promotes CD8 + T cell development by silencing Cd4 expression. Finally, studies have demonstrated the tumor suppressive activity of in a variety of cancers (e.g., gastric and breast cancers).
Description & Specifications

Specifications

Runx3
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
R3-5G4
AML2, CBFA3
Immunohistochemistry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL