missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RUNX3 Mouse anti-Human, eFluor® 660, Clone: R3-5G4, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS50981782

Catalog No. 501124556



Description

Description

Description: This R3-5G4 monoclonal antibody recognizes human Runx3, a 44 kDa transcription factor that belongs to the Runt domain-containing family. Runx3 is expressed in hematopoietic cells such as T cells, B cells, and myeloid cells. This transcription

This gene encodes a member of the runt domain-containing family of transcription factors. A heterodimer of this protein and a beta subunit forms a complex that binds to the core DNA sequence 5′-PYGPYGGT-3′ found in a number of enhancers and promoters, and can either activate or suppress transcription. It also interacts with other transcription factors. It functions as a tumor suppressor, and the gene is frequently deleted or transcriptionally silenced in cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Runx3
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q13761
AML2, CBFA3
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
R3-5G4
eFluor® 660
RUNX3
Liquid
RUNX3
IgG1, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
864
Documents

For Research Use Only.