missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rufinamide 98.0+%, TCI America™

Manufacturer:  Tci America R01431G

 View more versions of this product

Catalog No. R01431G



Specifications

Specifications

Rufinamide
241°C
106308-44-5
MFCD00865314
POGQSBRIGCQNEG-UHFFFAOYSA-N
1-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
129228
Crystal-Powder at 20°C
White
1g
C10H8F2N4O
rufinamide, inovelon, banzel, 1-2,6-difluorobenzyl-1h-1,2,3-triazole-4-carboxamide, xilep, 1-2,6-difluorophenyl methyl-1h-1,2,3-triazole-4-carboxamide, unii-wfw942pr79, 1-2,6-difluorophenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 1-2,6-difluorophenyl methyl, dsstox_cid_26506
C1=CC(=C(C(=C1)F)CN2C=C(N=N2)C(=O)N)F
238.198
238.20
≥98.0% (HPLC)

Chemical Identifiers

106308-44-5
238.198
POGQSBRIGCQNEG-UHFFFAOYSA-N
129228
C1=CC(=C(C(=C1)F)CN2C=C(N=N2)C(=O)N)F
C10H8F2N4O
MFCD00865314
rufinamide, inovelon, banzel, 1-2,6-difluorobenzyl-1h-1,2,3-triazole-4-carboxamide, xilep, 1-2,6-difluorophenyl methyl-1h-1,2,3-triazole-4-carboxamide, unii-wfw942pr79, 1-2,6-difluorophenyl methyl triazole-4-carboxamide, 1h-1,2,3-triazole-4-carboxamide, 1-2,6-difluorophenyl methyl, dsstox_cid_26506
1-[(2,6-difluorophenyl)methyl]triazole-4-carboxamide
Safety and Handling

Safety and Handling

TSCA : No

Documents