missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RuCl2[(S)-xylbinap][(S)-daipen], TCI America™

Manufacturer:  Tci America R01321G

 View more versions of this product

Catalog No. R01321G



Specifications

Specifications

RuCl2[(S)-xylbinap][(S)-daipen]
1g
C71H74Cl2N2O2P2Ru
1221.30
Undesignated
220114-01-2
MFCD09753027
Crystal-Powder at 20°C

Chemical Identifiers

220114-01-2
C71H74Cl2N2O2P2Ru
MFCD09753027
Safety and Handling

Safety and Handling

TSCA : No

Documents