missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
RuCl2[(S)-xylbinap][(S)-daipen], TCI America™
$132.71 - $449.59
Chemical Identifiers
CAS | 220114-01-2 |
---|---|
Molecular Formula | C71H74Cl2N2O2P2Ru |
MDL Number | MFCD09753027 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
R0132200MG
|
TCI America
R0132200MG |
200 mg |
Each for $132.71
|
|
|||||
R01321G
|
TCI America
R01321G |
1 g |
Each for $449.59
|
|
|||||
Chemical Identifiers
220114-01-2 | |
C71H74Cl2N2O2P2Ru | |
MFCD09753027 |
Specifications
220114-01-2 | |
MFCD09753027 | |
1221.30 | |
RuCl2[(S)-xylbinap][(S)-daipen] |
C71H74Cl2N2O2P2Ru | |
200 mg | |
Crystalline Powder |
Safety and Handling
TSCA : No