missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
RuCl2[(S)-dm-segphos(regR)][(S)-daipen], TCI America™
$112.81 - $393.19
Chemical Identifiers
CAS | 944450-44-6 |
---|---|
Molecular Formula | C65H70Cl2N2O6P2Ru |
MDL Number | MFCD09753029 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
R0128200MG
|
TCI America
R0128200MG |
200 mg |
Each for $112.81
|
|
|||||
R01281G
|
TCI America
R01281G |
1 g |
Each for $393.19
|
|
|||||
Chemical Identifiers
944450-44-6 | |
C65H70Cl2N2O6P2Ru | |
MFCD09753029 |
Specifications
944450-44-6 | |
MFCD09753029 | |
1209.20 | |
RuCl2[(S)-dm-segphos(regR)][(S)-daipen] |
C65H70Cl2N2O6P2Ru | |
200 mg | |
Crystalline Powder |
Safety and Handling
TSCA : No