missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RuCl2[(R)-xylbinap][(R,R)-dpen], TCI America™

$102.42 - $347.95

Chemical Identifiers

CAS 220114-38-5
Molecular Formula C66H64Cl2N2P2Ru
Molecular Weight (g/mol) 1119.17
MDL Number MFCD09753028
InChI Key HYMSONXJNGZZBM-ODQAEMFESA-L
Synonym Dichloro[(R)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl][(R,R)-1,2-diphenylethylenediamine]ruthenium(II)
PubChem CID 16220282
IUPAC Name [1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;(1R,2R)-1,2-diphenylethane-1,2-diamine
SMILES CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N.Cl[Ru]Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R0135200MG View Documents Tci America
R0135200MG
200mg Each for $102.42
 
R01351G View Documents Tci America
R01351G
1g Each for $347.95
 
Specifications

Chemical Identifiers

220114-38-5
1119.17
HYMSONXJNGZZBM-ODQAEMFESA-L
16220282
CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N.Cl[Ru]Cl
C66H64Cl2N2P2Ru
MFCD09753028
Dichloro[(R)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl][(R,R)-1,2-diphenylethylenediamine]ruthenium(II)
[1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;(1R,2R)-1,2-diphenylethane-1,2-diamine

Specifications

Undesignated
220114-38-5
MFCD09753028
HYMSONXJNGZZBM-ODQAEMFESA-L
[1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;(1R,2R)-1,2-diphenylethane-1,2-diamine
16220282
Crystal-Powder at 20°C
200mg
C66H64Cl2N2P2Ru
Dichloro[(R)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl][(R,R)-1,2-diphenylethylenediamine]ruthenium(II)
CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N.Cl[Ru]Cl
1119.17
1119.17
RuCl2[(R)-xylbinap][(R,R)-dpen]
Safety and Handling

Safety and Handling

TSCA : No