missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RuCl2[(R)-dm-segphos(regR)][(R,R)-dpen], TCI America™

Manufacturer:  Tci America R01311G

 View more versions of this product

Catalog No. R01311G



Specifications

Specifications

RuCl2[(R)-dm-segphos(regR)][(R,R)-dpen]
1g
C60H60Cl2N2O4P2Ru
FOEKPXQPRAIJRW-ODQAEMFESA-L
[4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;(1R,2R)-1,2-diphenylethane-1,2-diamine
71310561
Crystal-Powder at 20°C
Undesignated
944450-45-7
MFCD09753031
CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N.Cl[Ru]Cl
1107.068
1107.07

Chemical Identifiers

944450-45-7
1107.068
FOEKPXQPRAIJRW-ODQAEMFESA-L
[4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;(1R,2R)-1,2-diphenylethane-1,2-diamine
C60H60Cl2N2O4P2Ru
MFCD09753031
71310561
CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N.Cl[Ru]Cl
Safety and Handling

Safety and Handling

TSCA : No

Documents