missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RuCl2[(R)-dm-segphos(regR)][(R)-daipen], TCI America™

Manufacturer:  TCI America R01291G

 View more versions of this product

Catalog No. R01291G



Specifications

Specifications

RuCl2[(R)-dm-segphos(regR)][(R)-daipen]
1g
C65H70Cl2N2O6P2Ru
UDSABKQFPIPYOT-OEGAAENXSA-L
[4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;(2R)-1,1-bis(4-methoxyphenyl)-3-methylbutane-1,2-diamine;dichlororuthenium
71310545
Crystal-Powder at 20°C
Undesignated
944450-43-5
MFCD10567024
CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CC(C)C(C(C1=CC=C(C=C1)OC)(C2=CC=C(C=C2)OC)N)N.Cl[Ru]Cl
1209.201
1209.20

Chemical Identifiers

944450-43-5
1209.201
UDSABKQFPIPYOT-OEGAAENXSA-L
[4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;(2R)-1,1-bis(4-methoxyphenyl)-3-methylbutane-1,2-diamine;dichlororuthenium
C65H70Cl2N2O6P2Ru
MFCD10567024
71310545
CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CC(C)C(C(C1=CC=C(C=C1)OC)(C2=CC=C(C=C2)OC)N)N.Cl[Ru]Cl
Safety and Handling

Safety and Handling

TSCA : No

Documents