missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RuCl[(S,S)-Tsdpen](p-cymene) 90.0+%, TCI America™

Manufacturer:  Tci America R01241G

 View more versions of this product

Catalog No. R01241G



Specifications

Specifications

RuCl[(S,S)-Tsdpen](p-cymene)
1g
C31H35ClN2O2RuS
[(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
Crystal-Powder at 20°C
Undesignated
192139-90-5
MFCD09953484
636.21
≥90% (HPLC)

Chemical Identifiers

192139-90-5
C31H35ClN2O2RuS
MFCD09953484
[(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
Safety and Handling

Safety and Handling

TSCA : No

Documents