missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RuCl[(S,S)-Tsdpen](mesitylene) 90.0+%, TCI America™

Manufacturer:  Tci America R0126200MG

 View more versions of this product

Catalog No. R0126200MG



Specifications

Specifications

RuCl[(S,S)-Tsdpen](mesitylene)
200mg
C30H34ClN2O2RuS
chloro 1s,2s-n-p-toluenesulfonyl-1,2-diphenylethanediamine mesitylene ruthenium ii
CC1=CC=C(C=C1)S(=O)(=O)NC(C2=CC=CC=C2)C(C3=CC=CC=C3)N.CC1=CC(=CC(=C1)C)C.[Cl-].[Ru+]
623.194
622.19
≥90% (HPLC)
Undesignated
174813-81-1
MFCD12545952
BABSEZBJIPZYQB-XCPIVNJJSA-M
N-[(1S,2S)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide;ruthenium(1+);1,3,5-trimethylbenzene;chloride
131675785
Crystal-Powder at 20°C

Chemical Identifiers

174813-81-1
623.194
BABSEZBJIPZYQB-XCPIVNJJSA-M
131675785
CC1=CC=C(C=C1)S(=O)(=O)NC(C2=CC=CC=C2)C(C3=CC=CC=C3)N.CC1=CC(=CC(=C1)C)C.[Cl-].[Ru+]
C30H34ClN2O2RuS
MFCD12545952
chloro 1s,2s-n-p-toluenesulfonyl-1,2-diphenylethanediamine mesitylene ruthenium ii
N-[(1S,2S)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide;ruthenium(1+);1,3,5-trimethylbenzene;chloride
Safety and Handling

Safety and Handling

TSCA : No

Documents