missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RuCl[(S,S)-Fsdpen](p-cymene) 90.0+%, TCI America™

Manufacturer:  Tci America R0122200MG

 View more versions of this product

Catalog No. R0122200MG



Specifications

Specifications

RuCl[(S,S)-Fsdpen](p-cymene)
244°C
1026995-72-1
MFCD12545954
WILRHPOBOUTKKV-NWMPYMMKSA-M
N-[(1S,2S)-2-amino-1,2-diphenylethyl]-2,3,4,5,6-pentafluorobenzenesulfonamide;chlororuthenium(1+);1-methyl-4-propan-2-ylbenzene
132274302
Crystal-Powder at 20°C
Undesignated
200mg
C30H29ClF5N2O2RuS+
[(S,S)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)NS(=O)(=O)C3=C(C(=C(C(=C3F)F)F)F)F)N.Cl[Ru+]
713.146
712.14
≥90% (NMR)

Chemical Identifiers

1026995-72-1
713.146
WILRHPOBOUTKKV-NWMPYMMKSA-M
132274302
CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)NS(=O)(=O)C3=C(C(=C(C(=C3F)F)F)F)F)N.Cl[Ru+]
C30H29ClF5N2O2RuS+
MFCD12545954
[(S,S)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
N-[(1S,2S)-2-amino-1,2-diphenylethyl]-2,3,4,5,6-pentafluorobenzenesulfonamide;chlororuthenium(1+);1-methyl-4-propan-2-ylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents