missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RuCl[(R,R)-Tsdpen](p-cymene) 90.0+%, TCI America™

Manufacturer:  Tci America R0125200MG

 View more versions of this product

Catalog No. R0125200MG



Specifications

Specifications

RuCl[(R,R)-Tsdpen](p-cymene)
215°C
192139-92-7
MFCD10567385
636.21
≥90% (HPLC)
Undesignated
200mg
C31H35ClN2O2RuS
[(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(R,R)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
Crystal-Powder at 20°C

Chemical Identifiers

192139-92-7
C31H35ClN2O2RuS
MFCD10567385
[(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(R,R)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
Safety and Handling

Safety and Handling

TSCA : No

Documents