missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RuCl[(R,R)-Fsdpen](p-cymene) 90.0+%, TCI America™

Manufacturer:  Tci America R01231G

 View more versions of this product

Catalog No. R01231G



Specifications

Specifications

RuCl[(R,R)-Fsdpen](p-cymene)
244°C
1026995-71-0
MFCD12545963
UWFMZLATRGEOIW-ZJPTYAPPSA-M
[(1R,2R)-2-amino-1,2-diphenylethyl]-(2,3,4,5,6-pentafluorophenyl)sulfonylazanide;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);chloride
71463829
Crystal-Powder at 20°C
Undesignated
1g
C30H28ClF5N2O2RuS
[(R,R)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(R,R)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)[N-]S(=O)(=O)C3=C(C(=C(C(=C3F)F)F)F)F)N.[Cl-].[Ru+2]
712.138
712.14
≥90% (NMR)

Chemical Identifiers

1026995-71-0
712.138
UWFMZLATRGEOIW-ZJPTYAPPSA-M
71463829
CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)[N-]S(=O)(=O)C3=C(C(=C(C(=C3F)F)F)F)F)N.[Cl-].[Ru+2]
C30H28ClF5N2O2RuS
MFCD12545963
[(R,R)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(R,R)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
[(1R,2R)-2-amino-1,2-diphenylethyl]-(2,3,4,5,6-pentafluorophenyl)sulfonylazanide;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);chloride
Safety and Handling

Safety and Handling

TSCA : No

Documents