missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[RuCl(p-cymene)((S)-tolbinap)]Cl, TCI America™

Manufacturer:  Tci America R0149200MG

 View more versions of this product

Catalog No. R0149200MG



Specifications

Specifications

[RuCl(p-cymene)((S)-tolbinap)]Cl
200mg
C58H54Cl2P2Ru
Chloro[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl](p-cymene)ruthenium(II) Chloride
CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.[Cl-].[Cl-].[Ru+2]
984.988
984.99
Undesignated
228120-95-4
MFCD09753014
BHIUOOHVIWKUTN-UHFFFAOYSA-L
[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);dichloride
91972163
Crystal-Powder at 20°C

Chemical Identifiers

228120-95-4
984.988
BHIUOOHVIWKUTN-UHFFFAOYSA-L
91972163
CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.[Cl-].[Cl-].[Ru+2]
C58H54Cl2P2Ru
MFCD09753014
Chloro[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl](p-cymene)ruthenium(II) Chloride
[1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);dichloride
Safety and Handling

Safety and Handling

TSCA : No

Documents