missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[RuCl(p-cymene)((S)-dm-segphos(regR))]Cl, TCI America™

Manufacturer:  Tci America R01571G

 View more versions of this product

Catalog No. R01571G



Specifications

Specifications

[RuCl(p-cymene)((S)-dm-segphos(regR))]Cl
1g
C56H58Cl2O4P2Ru+
Chloro[(S)-(-)-5,5′C-bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
CC1=CC=C(C=C1)C(C)C.CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.[Cl-].[Cl-].[Ru+3]
1028.994
1028.99
Undesignated
944451-31-4
MFCD09753018
HXGKHUFSYGQNDT-UHFFFAOYSA-L
[4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(3+);dichloride
91972170
Crystal-Powder at 20°C

Chemical Identifiers

944451-31-4
1028.994
HXGKHUFSYGQNDT-UHFFFAOYSA-L
91972170
CC1=CC=C(C=C1)C(C)C.CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.[Cl-].[Cl-].[Ru+3]
C56H58Cl2O4P2Ru+
MFCD09753018
Chloro[(S)-(-)-5,5′C-bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
[4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(3+);dichloride
Safety and Handling

Safety and Handling

TSCA : No

Documents