missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
[RuCl(p-cymene)((S)-binap)]Cl 90.0+%, TCI America™
Supplier: TCI America R01471G
Specifications
[RuCl(p-cymene)((S)-binap)]Cl | |
C54H46Cl2P2Ru | |
1 g | |
WNHLGYRPKARUHY-UHFFFAOYSA-L | |
dichlororuthenium;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene | |
11147374 | |
≥90.0% (HPLC) |
130004-33-0 | |
MFCD00134456 | |
chloro s---2,2'-bis diphenylphosphino-1,1'-binaphthyl p-cymene ruthenium ii chloride, r-+-2,2-bis diphenylphosphino-1,1-binaphthalenechloro p-cymene ruthenium chloride, chloro r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl p-cymene ruthenium ii chloride, rucl p-cymene s-binap cl, r-2,2'-bis-diphenylphosphino-1,1'-binaphthyl-chloro-p-cymene-rutheniumchloride, s-2,2'-bis-diphenylphosphino-1,1'-binaphthyl-chloro-p-cymene-rutheniumchloride | |
CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl[Ru]Cl | |
928.88 | |
928.88 | |
Crystalline Powder |
Chemical Identifiers
130004-33-0 | |
928.88 | |
WNHLGYRPKARUHY-UHFFFAOYSA-L | |
11147374 | |
CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl[Ru]Cl |
C54H46Cl2P2Ru | |
MFCD00134456 | |
chloro s---2,2'-bis diphenylphosphino-1,1'-binaphthyl p-cymene ruthenium ii chloride, r-+-2,2-bis diphenylphosphino-1,1-binaphthalenechloro p-cymene ruthenium chloride, chloro r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl p-cymene ruthenium ii chloride, rucl p-cymene s-binap cl, r-2,2'-bis-diphenylphosphino-1,1'-binaphthyl-chloro-p-cymene-rutheniumchloride, s-2,2'-bis-diphenylphosphino-1,1'-binaphthyl-chloro-p-cymene-rutheniumchloride | |
dichlororuthenium;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene |
Safety and Handling
TSCA : No