missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[RuCl(p-cymene)((R)-xylbinap)]Cl, TCI America™

Manufacturer:  Tci America R01501G

 View more versions of this product

Catalog No. R01501G



Specifications

Specifications

[RuCl(p-cymene)((R)-xylbinap)]Cl
1g
C62H62Cl2P2Ru
Chloro[(R)-(+)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl](p-cymene)ruthenium(II) Chloride
CC1=CC=C(C=C1)C(C)C.CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.[Cl-].[Cl-].[Ru+2]
1041.096
1041.10
Undesignated
944451-24-5
MFCD09753015
CBZXDZCPCDMKJU-UHFFFAOYSA-L
[1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);dichloride
91972161
Crystal-Powder at 20°C

Chemical Identifiers

944451-24-5
1041.096
CBZXDZCPCDMKJU-UHFFFAOYSA-L
91972161
CC1=CC=C(C=C1)C(C)C.CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.[Cl-].[Cl-].[Ru+2]
C62H62Cl2P2Ru
MFCD09753015
Chloro[(R)-(+)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl](p-cymene)ruthenium(II) Chloride
[1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);dichloride
Safety and Handling

Safety and Handling

TSCA : No

Documents