missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[RuCl(p-cymene)((R)-dtbm-segphos(regR))]Cl, TCI America™

Manufacturer:  Tci America R01581G

 View more versions of this product

Catalog No. R01581G



Specifications

Specifications

[RuCl(p-cymene)((R)-dtbm-segphos(regR))]Cl
1g
C84H114Cl2O8P2Ru
Chloro[(R)-(-)-5,5′C-bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
CC1=CC=C(C=C1)C(C)C.CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)C7=CC(=C(C(=C7)C(C)(C)C)OC)C(C)(C)C)C8=CC(=C(C(=C8)C(C)(C)C)OC)C(C)(C)C.Cl[Ru]Cl
1485.746
1485.75
Undesignated
944451-32-5
MFCD09753019
MYYXUILRYSITRR-UHFFFAOYSA-L
[4-[5-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane;dichlororuthenium;1-methyl-4-propan-2-ylbenzene
71310560
Crystal-Powder at 20°C

Chemical Identifiers

944451-32-5
1485.746
MYYXUILRYSITRR-UHFFFAOYSA-L
71310560
CC1=CC=C(C=C1)C(C)C.CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)C7=CC(=C(C(=C7)C(C)(C)C)OC)C(C)(C)C)C8=CC(=C(C(=C8)C(C)(C)C)OC)C(C)(C)C.Cl[Ru]Cl
C84H114Cl2O8P2Ru
MFCD09753019
Chloro[(R)-(-)-5,5′C-bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
[4-[5-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane;dichlororuthenium;1-methyl-4-propan-2-ylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents