missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[RuCl(p-cymene)((R)-dtbm-segphos(regR))]Cl, TCI America™

$82.84 - $274.14

Chemical Identifiers

CAS 944451-32-5
Molecular Formula C84H114Cl2O8P2Ru
Molecular Weight (g/mol) 1485.746
MDL Number MFCD09753019
InChI Key MYYXUILRYSITRR-UHFFFAOYSA-L
Synonym Chloro[(R)-(-)-5,5′C-bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
PubChem CID 71310560
IUPAC Name [4-[5-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane;dichlororuthenium;1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C.CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)C7=CC(=C(C(=C7)C(C)(C)C)OC)C(C)(C)C)C8=CC(=C(C(=C8)C(C)(C)C)OC)C(C)(C)C.Cl[Ru]Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R01581G View Documents Tci America
R01581G
1g Each for $274.14
 
R0158200MG View Documents Tci America
R0158200MG
200mg Each for $82.84
 
Specifications

Chemical Identifiers

944451-32-5
1485.746
MYYXUILRYSITRR-UHFFFAOYSA-L
71310560
CC1=CC=C(C=C1)C(C)C.CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)C7=CC(=C(C(=C7)C(C)(C)C)OC)C(C)(C)C)C8=CC(=C(C(=C8)C(C)(C)C)OC)C(C)(C)C.Cl[Ru]Cl
C84H114Cl2O8P2Ru
MFCD09753019
Chloro[(R)-(-)-5,5′C-bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
[4-[5-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane;dichlororuthenium;1-methyl-4-propan-2-ylbenzene

Specifications

Undesignated
944451-32-5
MFCD09753019
MYYXUILRYSITRR-UHFFFAOYSA-L
[4-[5-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane;dichlororuthenium;1-methyl-4-propan-2-ylbenzene
71310560
Crystal-Powder at 20°C
1g
C84H114Cl2O8P2Ru
Chloro[(R)-(-)-5,5′C-bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
CC1=CC=C(C=C1)C(C)C.CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)C7=CC(=C(C(=C7)C(C)(C)C)OC)C(C)(C)C)C8=CC(=C(C(=C8)C(C)(C)C)OC)C(C)(C)C.Cl[Ru]Cl
1485.746
1485.75
[RuCl(p-cymene)((R)-dtbm-segphos(regR))]Cl
Safety and Handling

Safety and Handling

TSCA : No