missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[RuCl(p-cymene)((R)-binap)]Cl 90.0+%, TCI America™

Manufacturer:  Tci America R01461G

 View more versions of this product

Catalog No. R01461G



Specifications

Specifications

[RuCl(p-cymene)((R)-binap)]Cl
1g
C54H46Cl2P2Ru+
chloro r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl p-cymene ruthenium ii chloride, chloro r-+-2,2?-bis diphenylphosphino-1,1?-binaphthyl p-cymene ruthenium ii chloride, r-+-2,2-bis diphenylphosphino-1,1-binaphthalenechloro p-cymene ruthenium chloride, r-binap rucl p-cymene cl, rucl p-cymene r-binap cl
CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Cl-].[Cl-].[Ru+3]
928.88
928.88
≥90.0% (HPLC)
Undesignated
145926-28-9
MFCD00134456
MMAGOHNRINMYDK-UHFFFAOYSA-L
[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(3+);dichloride
91972169
Crystal-Powder at 20°C

Chemical Identifiers

145926-28-9
928.88
MMAGOHNRINMYDK-UHFFFAOYSA-L
91972169
CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Cl-].[Cl-].[Ru+3]
C54H46Cl2P2Ru+
MFCD00134456
chloro r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl p-cymene ruthenium ii chloride, chloro r-+-2,2?-bis diphenylphosphino-1,1?-binaphthyl p-cymene ruthenium ii chloride, r-+-2,2-bis diphenylphosphino-1,1-binaphthalenechloro p-cymene ruthenium chloride, r-binap rucl p-cymene cl, rucl p-cymene r-binap cl
[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(3+);dichloride
Safety and Handling

Safety and Handling

TSCA : No

Documents