missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubrene, 97%

Manufacturer:  Alfa Aesar™ H5350706

 View more versions of this product

Catalog No. AAH5350706



Description

Description

Applications
Rubrene is an organic semiconductor used in organic light-emitting diodes (OLEDs) and organic field-effect transistors. It is used as a sensitizer in chemoluminesence. It is also used to prepare single crystal transistors. It acts as a reagent for chemiluminescence research and for transition metal complex ligation. Further, it is used as a p-type organic semiconductor.

Solubility
Soluble in hot toluene. Insoluble in water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Rubrene
330°C to 335°C
517-51-1
MFCD00003703
5,11,12-tetraphenylnaphthacene
YYMBJDOZVAITBP-UHFFFAOYSA-N
5,6,11,12-tetraphenyltetracene
68203
97%
Air Sensitive
5g
C42H28
1917339
Soluble in hot toluene. Insoluble in water.
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8
532.686
532.68

Chemical Identifiers

517-51-1
532.686
YYMBJDOZVAITBP-UHFFFAOYSA-N
68203
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8
C42H28
MFCD00003703
5,11,12-tetraphenylnaphthacene
5,6,11,12-tetraphenyltetracene
Safety and Handling

Safety and Handling

EINECSNumber : 208-242-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents