Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubidium sulfate, Puratronic™, 99.975% (metals basis)

Chemical Identifiers

Chemical Name or Material Rubidium sulfate
CAS 7488-54-2
Purity Grade Puratronic™
Formula Weight 267
MDL Number MFCD00011190
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1287204

  SDS
alfa aesar™
1287204
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1287209

  SDS
alfa aesar™
1287209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 267
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Assay Percent Notes (metals basis)
Refractive Index 1.513
Odor Odorless
CAS 7488-54-2
Assay 99.975%
Quantity 2g
Chemical Name or Material Rubidium sulfate
TSCA TSCA
MDL Number MFCD00011190
Physical Form Crystalline
Melting Point 1050°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Purity Grade Puratronic™
Density 3.613g/mL
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.