missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rubidium sulfate, 99.8% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01056709
Description
Rubidium sulfate is used in hematology stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Rubidium sulfate | |
7488-54-2 | |
10 g | |
H2O4RbS | |
rubidium sulfate, rubidium sulphate, dirubidium sulfate, rubidium sulfate rb2so4, sulfuric acid, dirubidium salt, rubidium sulfate rb2 so4, dirubidium 1+ sulfate, rubidiumsulfat, sulfuric acid, rubidium salt 1:2, rubidium 1+ sulfate | |
XNNUAZGCOYTNQE-UHFFFAOYSA-N | |
rubidium(1+);sulfate | |
197088 | |
99.8% | |
1.513 |
1050°C | |
Powder | |
(metals basis) | |
MFCD00011190 | |
Soluble in water. | |
[Rb].OS(O)(=O)=O | |
183.54 | |
267 | |
Odorless | |
3.613 g/mL |
Chemical Identifiers
7488-54-2 | |
183.54 | |
XNNUAZGCOYTNQE-UHFFFAOYSA-N | |
[Rb].OS(O)(=O)=O |
H2O4RbS | |
MFCD00011190 | |
197088 |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 231-301-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only