missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubidium sulfate, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 1056718

 View more versions of this product

Catalog No. AA1056718



Specifications

Specifications

Rubidium sulfate
(metals basis)
MFCD00011190
GANPIEKBSASAOC-UHFFFAOYSA-L
rubidium(1+);sulfate
197088
Powder
3.613g/mL
Odorless
1050°C
7488-54-2
O4Rb2S
rubidium sulfate, rubidium sulphate, dirubidium sulfate, rubidium sulfate rb2so4, sulfuric acid, dirubidium salt, rubidium sulfate rb2 so4, dirubidium 1+ sulfate, rubidiumsulfat, sulfuric acid, rubidium salt 1:2, rubidium 1+ sulfate
[O-]S(=O)(=O)[O-].[Rb+].[Rb+]
266.992
267
99.8%
−20 Mesh
1.513
50g

Chemical Identifiers

7488-54-2
266.992
GANPIEKBSASAOC-UHFFFAOYSA-L
197088
[O-]S(=O)(=O)[O-].[Rb+].[Rb+]
O4Rb2S
MFCD00011190
rubidium sulfate, rubidium sulphate, dirubidium sulfate, rubidium sulfate rb2so4, sulfuric acid, dirubidium salt, rubidium sulfate rb2 so4, dirubidium 1+ sulfate, rubidiumsulfat, sulfuric acid, rubidium salt 1:2, rubidium 1+ sulfate
rubidium(1+);sulfate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

Documents