missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium, plasma standard solution, Rb 1000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13872AE

 View more versions of this product

Catalog No. AA13872AE



Specifications

Specifications

Rubidium
Clear liquid
MFCD00134055
WPFGFHJALYCVMO-UHFFFAOYSA-L
rubidium(1+);carbonate
11431
23 hPa (17mm Hg) at 20°C
100mL
584-09-8,7697-37-2,7732-18-5
CO3Rb2
Nitric acid: 5.0%; Rubidium carbonate: 0.14%; Water: 94.86%
C(=O)([O-])[O-].[Rb+].[Rb+]
230.944
Liquid
Colorless

Chemical Identifiers

584-09-8
230.944
WPFGFHJALYCVMO-UHFFFAOYSA-L
rubidium(1+);carbonate
CO3Rb2
MFCD00134055
11431
C(=O)([O-])[O-].[Rb+].[Rb+]
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Documents