Learn More
Alfa Aesar™ Rubidium nitrate, 99% (metals basis)
As a catalyst in organic reactions and as chelating agents
Manufacturer: Alfa Aesar™ 1349614
Specifications
Rubidium nitrate | |
(metals basis) | |
MFCD00011193 | |
rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate | |
RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
rubidium(1+);nitrate | |
25731 | |
Crystalline | |
3.11g/mL | |
Hygroscopic | |
310°C |
13126-12-0 | |
NO3Rb | |
UN1477 | |
Soluble in water; Slightly soluble in acetone; Very soluble in HNO3 | |
[N+](=O)([O-])[O-].[Rb+] | |
147.472 | |
147.47 | |
99% | |
1.52 | |
White | |
25g |
Chemical Identifiers
13126-12-0 | |
147.472 | |
RTHYXYOJKHGZJT-UHFFFAOYSA-N | |
25731 | |
[N+](=O)([O-])[O-].[Rb+] |
NO3Rb | |
MFCD00011193 | |
rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate | |
rubidium(1+);nitrate |
Safety and Handling
GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
RTECSNumber : QV0900000
TSCA : TSCA