Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubidium nitrate, 99% (metals basis)

As a catalyst in organic reactions and as chelating agents

Chemical Identifiers

Chemical Name or Material Rubidium nitrate
CAS 13126-12-0
Formula Weight 147.47
MDL Number MFCD00011193
UN Number UN1477
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1349614

  SDS
alfa aesar™
1349614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1349622

  SDS
alfa aesar™
1349622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.47
Health Hazard 1 Danger
Refractive Index 1.52
Color White
Sensitivity Hygroscopic
Density 3.11g/mL
RTECS Number QV0900000
CAS 13126-12-0
Solubility Information Soluble in water; Slightly soluble in acetone; Very soluble in HNO3
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
Physical Form Crystalline
Melting Point 310°C
Chemical Name or Material Rubidium nitrate
Health Hazard 3 GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN1477
MDL Number MFCD00011193
Health Hazard 2 GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
TSCA TSCA
Assay 99%
Assay Percent Notes (metals basis)
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.