missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium molybdenum oxide, 99.5% (Metals basis), Alfa Aesar™

Chemical Identifiers

CAS 13718-22-4
Physical Form Powder
Assay Percent Range 99.5% (metals basis)
Molecular Formula Rb2MoO4
Formula Weight 330.88
MDL Number MFCD00049518
Synonym Rubidium molybdate
Chemical Name or Material Rubidium molybdenum oxide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3951604 SDS Alfa Aesar
3951604
2g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA3951609 SDS Alfa Aesar
3951609
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA3951618 SDS Alfa Aesar
3951618
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

13718-22-4
99.5% (metals basis)
330.88
Rubidium molybdate
Powder
Rb2MoO4
MFCD00049518
Rubidium molybdenum oxide

Specifications

330.88
2g
Rubidium molybdate
Odorless
13718-22-4
MFCD00049518
Powder
White
Rb2MoO4
99.5% (metals basis)
White powder
Rubidium molybdenum oxide

Safety and Handling

H373-H319-H335: May cause damage to organs through prolonged or repeated exposure. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 237-271-1