Please login to your online account to display your discounted pricing

Rubidium iodide, 99.9%, extra pure, Acros Organics

Chemical Identifiers

Chemical Name or Material Rubidium iodide
Name Note Extra Pure
CAS 7790-29-6
Purity Grade Extra Pure
Assay Percent Range 99.9%
Molecular Formula IRb
Linear Formula RbI
Formula Weight 212.37
MDL Number MFCD00011192
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC318420100

  SDS
thermo fisher scientific
AC318420100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.37
Physical Form Crystalline Powder
Purity Grade Extra Pure
Quantity 10g
Color White
Molecular Formula IRb
CAS 7790-29-6
Name Note Extra Pure
Chemical Name or Material Rubidium iodide
Assay Percent Range 99.9%
MDL Number MFCD00011192
Linear Formula RbI
Packaging Glass bottle
Total Trace Metal Impurities 0.1% max.