Please login to your online account to display your discounted pricing

Rubidium iodide, 99.9%, extra pure, Acros Organics


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC318420100

 View more versions of this product

Catalog No. AC318420100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Crystalline Powder
Formula Weight 212.37
Quantity 10g
Color White
Chemical Name or Material Rubidium iodide
Name Note Extra Pure
Packaging Glass bottle
Assay Percent Range 99.9%
Molecular Formula IRb
Linear Formula RbI
MDL Number MFCD00011192
CAS 7790-29-6
Total Trace Metal Impurities 0.1% max.

Specifications

Chemical Name or Material Rubidium iodide
Name Note Extra Pure
CAS 7790-29-6
Purity Grade Extra Pure
Assay Percent Range 99.9%
Molecular Formula IRb
Linear Formula RbI
Formula Weight 212.37
MDL Number MFCD00011192