missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium iodide, 99.8% (metals basis), Alfa Aesar™

$75.88 - $75.88

Chemical Identifiers

CAS 7790-29-6
Physical Form Crystals
Assay Percent Range 99.8% (metals basis)
Molecular Formula RbI
Formula Weight 212.37
MDL Number MFCD00011192
Chemical Name or Material Rubidium iodide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1349709 SDS Alfa Aesar
1349709
10g Each for $75.88
AA1349718 SDS Alfa Aesar
1349718
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7790-29-6
99.8% (metals basis)
212.37
Rubidium iodide
Crystals
RbI
MFCD00011192

Specifications

Crystals
10g
3.55g/cm3
Odorless
99.8% (metals basis)
MFCD00011192
212.37
White
RbI
7790-29-6
White crystalline
Rubidium iodide

Safety and Handling

EINECSNumber : 232-198-1