missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubidium iodide, 99.8% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1349718

 View more versions of this product

Catalog No. AA1349718



Specifications

Specifications

Rubidium iodide
99.8% (metals basis)
MFCD00011192
rubidium iodide, rubidium iodide, rubidium-iodid, rubidium jodatum, unii-18cyw0vl2x, rubidium 1+ iodide, rubidium 1+ ion iodide, 18cyw0vl2x, i.rb, acmc-20ajs7
[Rb+].[I-]
212.372
212.37
Odorless
50g
7790-29-6
IRb
White crystalline
WFUBYPSJBBQSOU-UHFFFAOYSA-M
rubidium(1+);iodide
3423208
Crystals
White

Chemical Identifiers

7790-29-6
212.372
WFUBYPSJBBQSOU-UHFFFAOYSA-M
3423208
[Rb+].[I-]
IRb
MFCD00011192
rubidium iodide, rubidium iodide, rubidium-iodid, rubidium jodatum, unii-18cyw0vl2x, rubidium 1+ iodide, rubidium 1+ ion iodide, 18cyw0vl2x, i.rb, acmc-20ajs7
rubidium(1+);iodide
Safety and Handling

Safety and Handling

EINECSNumber : 232-198-1

Documents