missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis)

Manufacturer:  Alfa Aesar™ 10565NE

 View more versions of this product

Catalog No. AA10565NE



Specifications

Specifications

Rubidium hydroxide
(metals basis)
RbOH
UN2677
rb.ho
[OH-].[Rb+]
102.475
CHEBI:32108
Liquid
50% w/w aq. soln.
50g
1310-82-3
HORb
MFCD00011194
14,8285
CPRMKOQKXYSDML-UHFFFAOYSA-M
rubidium(1+);hydroxide
62393
102.48 (Anhydrous)
≥99.6%
Yellow

Chemical Identifiers

1310-82-3
102.475
CPRMKOQKXYSDML-UHFFFAOYSA-M
62393
rubidium(1+);hydroxide
HORb
MFCD00011194
rb.ho
CHEBI:32108
[OH-].[Rb+]
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: RUBIDIUM HYDROXIDE SOLUTION

RTECSNumber : VL8750000

TSCA : TSCA

Documents