missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubidium fluoride, 99.1% (metals basis)

Manufacturer:  Alfa Aesar™ 4714418

 View more versions of this product

Catalog No. AA4714418



Specifications

Specifications

Rubidium fluoride
(metals basis)
RbF
UN3288
AHLATJUETSFVIM-UHFFFAOYSA-M
rubidium(1+);fluoride
83473
Crystalline
3.557g/mL
Hygroscopic
795°C
13446-74-7
FRb
MFCD00011191
rubidium fluoride, rubidium fluoride rbf, rubidium monofluoride, rubidium 1+ fluoride, rubidium 1+ ion fluoride, rubidium fluoride, anhydrous, unii-63564tnj4r, acmc-20ajsd, rubidium i fluoride, rubidium fluoride, rbf
[F-].[Rb+]
104.466
104.47
99.1%
1.398
White
50g

Chemical Identifiers

13446-74-7
104.466
AHLATJUETSFVIM-UHFFFAOYSA-M
83473
[F-].[Rb+]
FRb
MFCD00011191
rubidium fluoride, rubidium fluoride rbf, rubidium monofluoride, rubidium 1+ fluoride, rubidium 1+ ion fluoride, rubidium fluoride, anhydrous, unii-63564tnj4r, acmc-20ajsd, rubidium i fluoride, rubidium fluoride, rbf
rubidium(1+);fluoride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : VL8740000

TSCA : TSCA

SDS Documents