Please login to your online account to display your discounted pricing

Alfa Aesar™ Rubidium chloride, 99% (metals basis)

Testing for perchloric acid, source of rubidium, catalyst

$213.96 - $213.96

Chemical Identifiers

Chemical Name or Material Rubidium chloride
CAS 7791-11-9
Molecular Formula RbCl
Formula Weight 120.92
MDL Number MFCD00011187
Merck Index 14,8284
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1289206

  SDS
alfa aesar™
1289206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1289222

  SDS
alfa aesar™
1289222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1289214

  SDS
alfa aesar
1289214
25g Each for $213.96
Description & Specifications

Specifications

Formula Weight 120.92
Assay Percent Range 99% (metals basis)
Molecular Formula RbCl
Sensitivity Hygroscopic
Solubility Information Soluble in water; Practically insoluble in alcohol
Physical Form Crystalline
Boiling Point 1390°C
Color White
Assay 99%
Quantity 5g
MDL Number MFCD00011187
Melting Point 718°C
RTECS Number VL8575000
Odor Odorless
Chemical Name or Material Rubidium chloride
Assay Percent Notes (metals basis)
CAS 7791-11-9
Density 2.800g/mL
TSCA TSCA
Refractive Index 1.493
Merck Index 14,8284